D-gamma-Tocopherol, 95%
D-gamma-Tocopherol, 95%
D-gamma-Tocopherol, 95%
D-gamma-Tocopherol, 95%
Thermo Scientific Chemicals
D-gamma-Tocopherol, 95%
CAS: 54-28-4 | C28H48O2 | 416.69 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA421041000100 mgGlass bottle
Catalog number FSA421041000
Price (MYR)
1,456.00
Add to cart
Quantity:
100 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,456.00
Add to cart
Chemical Identifiers
CAS80-63-7
IUPAC Namemethyl 2-chloroprop-2-enoate
Molecular FormulaC4H5ClO2
InChI KeyAWJZTPWDQYFQPQ-UHFFFAOYSA-N
SMILESCOC(=O)C(Cl)=C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Infrared spectrumConforms
GC>=98.0 %
Refractive index1.4420 to 1.4440 (20°C, 589 nm)
StabilizerAbout 0.06% Hydroquinone