Tannic acid, ACS reagent, Thermo Scientific Chemicals
Tannic acid, ACS reagent, Thermo Scientific Chemicals
Tannic acid, ACS reagent, Thermo Scientific Chemicals
Tannic acid, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Tannic acid, ACS reagent, Thermo Scientific Chemicals
CAS: 1401-55-4 | C76H52O46 | 1701.206 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
Packaging:
Plastic bottle
Catalog number FSA419995000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS627-75-8
IUPAC Namehydrogen (2R)-2-amino-5-[(diaminomethylidene)amino]pentanoic acid chloride
Molecular FormulaC6H15ClN4O2
InChI KeyKWTQSFXGGICVPE-PGMHMLKASA-N
SMILES[H+].[Cl-].N[C@H](CCCN=C(N)N)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Optical Rotation-22? ? 1.5? (c=2 in 6M HCl)
Assay (Titration ex Chloride)≥97.5 to ≤102.5%
Appearance (Color)White to pale cream