Sulfuric acid, extra pure, fuming, 20-30% free SO3, Thermo Scientific Chemicals
Sulfuric acid, extra pure, fuming, 20-30% free SO3, Thermo Scientific Chemicals
Sulfuric acid, extra pure, fuming, 20-30% free SO3, Thermo Scientific Chemicals
Sulfuric acid, extra pure, fuming, 20-30% free SO3, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Sulfuric acid, extra pure, fuming, 20-30% free SO3, Thermo Scientific Chemicals
CAS: 8014-95-7 | H2O7S2 | 178.13 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA419975000500 gGlass bottle
Catalog number FSA419975000
Price (MYR)
912.00
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
912.00
Add to cart
Chemical Identifiers
CAS873-77-8
IUPAC Namebromo(4-chlorophenyl)magnesium
Molecular FormulaC6H4BrClMg
InChI KeyCDEMHJCJMMOFMB-UHFFFAOYSA-M
SMILESClC1=CC=C([Mg]Br)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Titration)0.8-1.2 M
Appearance (Color)Clear to slightly hazy