Stoddard Solvent, Thermo Scientific Chemicals
Stoddard Solvent, Thermo Scientific Chemicals
Stoddard Solvent, Thermo Scientific Chemicals
Stoddard Solvent, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Stoddard Solvent, Thermo Scientific Chemicals
CAS: 8052-41-3 | C8H15BrO2 | 223.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5 kg
Catalog number FSA419730010
Price (MYR)
580.00
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (MYR)
580.00
Add to cart
Chemical Identifiers
CAS110-82-7
IUPAC Namecyclohexane
Molecular FormulaC6H12
InChI KeyXDTMQSROBMDMFD-UHFFFAOYSA-N
SMILESC1CCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Water=<0.01 %
UVat 200 nm A: =<0.3
UVat 220 nm A: =<0.1
UVat 240 nm A: =<0.005
UV1 cm cell vs water