Stearic anhydride, 98%
Stearic anhydride, 98%
Stearic anhydride, 98%
Stearic anhydride, 98%
Thermo Scientific Chemicals
Stearic anhydride, 98%
CAS: 638-08-4 | C36H70O3 | 550.95 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA41971025025 gGlass bottle
Catalog number FSA419710250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialStearic anhydride
Name Note98%
CAS638-08-4
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement