Sodium biphenyl, 15-25%w/w solution in diethylene glycol diethyl ether, offered as 20 x 15mL
Sodium biphenyl, 15-25%w/w solution in diethylene glycol diethyl ether, offered as 20 x 15mL
Sodium biphenyl, 15-25%w/w solution in diethylene glycol diethyl ether, offered as 20 x 15mL
Sodium biphenyl, 15-25%w/w solution in diethylene glycol diethyl ether, offered as 20 x 15mL
Alfa Aesar
Sodium biphenyl, 15-25%w/w solution in diethylene glycol diethyl ether, offered as 20 x 15mL
CAS: 5137-46-2 | C12H10Na | 177.202 g/mol
Have Questions?
Catalog NumberQuantity
FSA419423000300 mL
Catalog number FSA419423000
Price (MYR)
1,617.00
Add to cart
Quantity:
300 mL
Request bulk or custom format
Price (MYR)
1,617.00
Add to cart
Chemical Identifiers
CAS412947-54-7
IUPAC Namemethyl 3-amino-4-iodobenzoate
Molecular FormulaC8H8INO2
InChI KeyWJEBNIVVLJEIKE-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC(N)=C(I)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
Melting Point (clear melt)130.0-140.0?C
FormPowder
Appearance (Color)Yellow
Reduction reagent
RUO – Research Use Only