Silver phosphate, 98%
Silver phosphate, 98%
Silver phosphate, 98%
Silver phosphate, 98%
Thermo Scientific Chemicals
Silver phosphate, 98%
CAS: 7784-09-0 | Ag3O4P | 418.57 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA41937025025 gGlass bottle
Catalog number FSA419370250
Price (MYR)
829.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
829.00
Add to cart
Specifications
Chemical Name or MaterialSilver phosphate
Melting Point849°C
Name Note98%
CAS7784-09-0
ColorGreen-Yellow to Yellow