Diethyldithiocarbamic acid, silver salt, ACS reagent, Thermo Scientific Chemicals
Diethyldithiocarbamic acid, silver salt, ACS reagent, Thermo Scientific Chemicals
Diethyldithiocarbamic acid, silver salt, ACS reagent, Thermo Scientific Chemicals
Diethyldithiocarbamic acid, silver salt, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Diethyldithiocarbamic acid, silver salt, ACS reagent, Thermo Scientific Chemicals
CAS: 1470-61-7 | C5H10AgNS2 | 256.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
25 g
Packaging:
Glass bottle
Catalog number FSA419330100
Price (MYR)
1,276.00
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,276.00
Add to cart
Chemical Identifiers
CAS109-00-2
IUPAC Namepyridin-3-ol
Molecular FormulaC5H5NO
InChI KeyGRFNBEZIAWKNCO-UHFFFAOYSA-N
SMILESOC1=CC=CN=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Water=<0.2 %
Appearance (Color)Beige to brown
Melting point123°C to 130°C
Appearance (Form)Crystalline powder