Rhodamine 101 inner salt, 94%, pure
Rhodamine 101 inner salt, 94%, pure
Rhodamine 101 inner salt, 94%, pure
Rhodamine 101 inner salt, 94%, pure
Thermo Scientific Chemicals
Rhodamine 101 inner salt, 94%, pure
CAS: 116450-56-7 | C32H30N2O3 | 490.603 g/mol
Have Questions?
Catalog NumberQuantity
FSA4190600101 g
Catalog number FSA419060010
Price (MYR)
1,424.00
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
1,424.00
Add to cart
Specifications
Chemical Name or MaterialRhodamine 101 inner salt
Melting Point271.0°C
Name Note94%
CAS116450-56-7
Health Hazard 2GHS H Statement: