Rhodamine B, 99+%, pure, laser grade
Rhodamine B, 99+%, pure, laser grade
Rhodamine B, 99+%, pure, laser grade
Rhodamine B, 99+%, pure, laser grade
Thermo Scientific Chemicals
Rhodamine B, 99+%, pure, laser grade
CAS: 81-88-9 | C28H31ClN2O3 | 479.02 g/mol
Have Questions?
Catalog NumberQuantity
FSA4190000101 g
Catalog number FSA419000010
Price (MYR)
681.00
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
681.00
Add to cart
Specifications
Chemical Name or MaterialRhodamine B
Melting Point210.0°C to 211.0°C
Name Note99+%
CAS81-88-9
Health Hazard 1Danger