Pyridine, 99+%, ACS reagent, Thermo Scientific Chemicals
Pyridine, 99+%, ACS reagent, Thermo Scientific Chemicals
Pyridine, 99+%, ACS reagent, Thermo Scientific Chemicals
Pyridine, 99+%, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Pyridine, 99+%, ACS reagent, Thermo Scientific Chemicals
CAS: 110-86-1 | C5H5N | 79.102 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
2.5 L
Packaging:
View all
EcoSafPak™ Glass Bottle
Catalog number FSA418545000
Price (MYR)
636.00
Add to cart
Quantity:
500 mL
Packaging:
EcoSafPak™ Glass Bottle
Request bulk or custom format
Price (MYR)
636.00
Add to cart
Chemical Identifiers
CAS6104-59-2
IUPAC Namesodium {4-[(4-ethoxyphenyl)amino]phenyl}bis(4-{ethyl[(3-sulfonatophenyl)methyl]amino}phenyl)methylium
Molecular FormulaC45H44N3NaO7S2
InChI KeyHAEGGLCXLFWTBE-UHFFFAOYSA-M
SMILES[Na+].CCOC1=CC=C(NC2=CC=C(C=C2)[C+](C2=CC=C(C=C2)N(CC)CC2=CC=CC(=C2)S([O-])(=O)=O)C2=CC=C(C=C2)N(CC)CC2=CC=CC(=C2)S([O-])(=O)=O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Bordeaux to dark purple-brown
Infrared spectrumConforms
Appearance (Form)Crystalline powder
Absorption Ratio P-15/P+151 to 1.2
UV Lambda max588 to 596 nm (in 50:50 ethanol:water)