Pyridine, 99+%, ACS reagent, Thermo Scientific Chemicals
Pyridine, 99+%, ACS reagent, Thermo Scientific Chemicals
Pyridine, 99+%, ACS reagent, Thermo Scientific Chemicals
Pyridine, 99+%, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Pyridine, 99+%, ACS reagent, Thermo Scientific Chemicals
CAS: 110-86-1 | C5H5N | 79.102 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
500 mL
1 L
Packaging:
View all
Glass Bottle
Catalog number FSA418540025
Price (MYR)
1,529.00
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
1,529.00
Add to cart
Specifications
Chemical Name or MaterialPyridine
Melting Point-42.0°C
CAS110-86-1
ColorColorless
Health Hazard 1Danger