Pyridine, 99+%, ACS reagent, Thermo Scientific Chemicals
Pyridine, 99+%, ACS reagent, Thermo Scientific Chemicals
Pyridine, 99+%, ACS reagent, Thermo Scientific Chemicals
Pyridine, 99+%, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Pyridine, 99+%, ACS reagent, Thermo Scientific Chemicals
CAS: 110-86-1 | C5H5N | 79.102 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
500 mL
2.5 L
Packaging:
View all
Glass Bottle
Catalog number FSA418540010
Price (MYR)
926.00
Add to cart
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
926.00
Add to cart
Chemical Identifiers
CAS7440-57-5
IUPAC Namegold
Molecular FormulaAu
InChI KeyPCHJSUWPFVWCPO-UHFFFAOYSA-N
SMILES[Au]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA≥99.9% (metals basis)
Appearance (Color)Gold
FormFoil, 1.0 micron thick.