2-Propene-1-sulfonic acid sodium salt, 95%, pract.
2-Propene-1-sulfonic acid sodium salt, 95%, pract.
2-Propene-1-sulfonic acid sodium salt, 95%, pract.
2-Propene-1-sulfonic acid sodium salt, 95%, pract.
Thermo Scientific Chemicals
2-Propene-1-sulfonic acid sodium salt, 95%, pract.
CAS: 2495-39-8 | C3H5NaO3S | 144.13 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA41837025025 gGlass bottle
Catalog number FSA418370250
Price (MYR)
614.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
614.00
Add to cart
Chemical Identifiers
CAS7790-69-4
IUPAC Namelithium(1+) nitrate
Molecular FormulaLiNO3
InChI KeyIIPYXGDZVMZOAP-UHFFFAOYSA-N
SMILES[Li+].[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder and/or granules
Assay (special)?2.5 % of Theoretical value (Li)
Total Metal Impurities≤0.001% (10 ppm)
Appearance (Color)White