Phosphomolybdic acid hydrate, ACS reagent, Thermo Scientific Chemicals
Phosphomolybdic acid hydrate, ACS reagent, Thermo Scientific Chemicals
Phosphomolybdic acid hydrate, ACS reagent, Thermo Scientific Chemicals
Phosphomolybdic acid hydrate, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Phosphomolybdic acid hydrate, ACS reagent, Thermo Scientific Chemicals
CAS: 51429-74-4 | H3Mo12O40P | 1825.40 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
Packaging:
View all
Glass Bottle
Catalog number FSA417891000
Price (MYR)
936.00
Add to cart
Quantity:
100 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
936.00
Add to cart
Specifications
Chemical Name or MaterialPhosphomolybdic Acid hydrate
ColorYellow
CAS51429-74-4
Health Hazard 1Danger
Health Hazard 2GHS H Statement:
Causes severe skin burns and eye damage.
May intensify fire; oxidizer.
View more
Oxidation reagent
RUO – Research Use Only