Petrolatum, white, pure, Thermo Scientific Chemicals
Petrolatum, white, pure, Thermo Scientific Chemicals
Petrolatum, white, pure, Thermo Scientific Chemicals
Petrolatum, white, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Petrolatum, white, pure, Thermo Scientific Chemicals
CAS: 8009-03-8 | C15H15N | 209.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
Packaging:
Glass bottle
Catalog number FSA417090010
Price (MYR)
328.00
Add to cart
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
328.00
Add to cart
Chemical Identifiers
CAS582-17-2
IUPAC Namenaphthalene-2,7-diol
Molecular FormulaC10H8O2
InChI KeyDFQICHCWIIJABH-UHFFFAOYSA-N
SMILESOC1=CC2=CC(O)=CC=C2C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale grey or pale brown
FormCrystals or powder or crystalline powder
Assay (Silylated GC)≥96.0%
Assay (HPLC)≥96.0%
Melting Point (clear melt)184-191?C