Pentafluoropropionic acid, 97%, Thermo Scientific Chemicals
Pentafluoropropionic acid, 97%, Thermo Scientific Chemicals
Pentafluoropropionic acid, 97%, Thermo Scientific Chemicals
Pentafluoropropionic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Pentafluoropropionic acid, 97%, Thermo Scientific Chemicals
CAS: 422-64-0 | C3HF5O2 | 164.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Packaging:
Plastic bottle
Catalog number FSA416920100
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS123-76-2
IUPAC Name4-oxopentanoic acid
Molecular FormulaC5H8O3
InChI KeyJOOXCMJARBKPKM-UHFFFAOYSA-N
SMILESCC(=O)CCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
AppearanceMay darken on storage
Titration with NaOH>=98.0 %
Appearance (Color)Colorless to white to yellow to orange
Refractive index1.4390 to 1.4430 (20°C, 589 nm)
Appearance (Form)Liquid after melting or solid