Paraformaldehyde, 96%, Thermo Scientific Chemicals
Paraformaldehyde, 96%, Thermo Scientific Chemicals
Paraformaldehyde, 96%, Thermo Scientific Chemicals
Paraformaldehyde, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Paraformaldehyde, 96%, Thermo Scientific Chemicals
Paraformaldehyde, 96%, CH2O, CAS Number-30525-89-4, paraformaldehyde, paraform, oxymethylene, methylene oxide, methyl aldehyde, methanal, formol, formalin, formic aldehyde, oxomethane, 500g, CHEBI:16842, White, 260 deg.C, 1.03 (Air - 1.0), 71 deg.C, Extra Pure, Warning
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
25 g
500 g
Packaging:
View all
Plastic bottle
Catalog number FSA416780010
Price (MYR)
490.00
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
490.00
Add to cart
Chemical Identifiers
CAS10450-60-9
IUPAC Nametetrahydroxyiodous acid
Molecular FormulaH5IO6
InChI KeyTWLXDPFBEPBAQB-UHFFFAOYSA-N
SMILESO[I](O)(O)(O)(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Sodium (Na)=<100 ppm
Zinc (Zn)=<50 ppm
Nitrate (NO3)=<100 ppm
Sulfate (SO4)=<100 ppm
Chloride and Chlorate (as Cl)=<200 ppm