Paraffin, pure, wax, granular, Thermo Scientific Chemicals
Paraffin, pure, wax, granular, Thermo Scientific Chemicals
Paraffin, pure, wax, granular, Thermo Scientific Chemicals
Paraffin, pure, wax, granular, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Paraffin, pure, wax, granular, Thermo Scientific Chemicals
Paraffin wax, CnH2n+2, CAS Number-8002-74-2, propafenona inn-spanish, propafenona, propafenone-hcl, propafenonum inn-latin, propafenonum, propafenone inn:ban, propafenone inn, rythmol, propafenone, gnf-pf-4594, 2kg, 322.0 deg.C, CHEBI:63619, White, 232-315-6, 198 deg.C, Pure
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 kg
10 kg
Catalog number FSA416770020
Price (MYR)
688.00
Add to cart
Quantity:
2 kg
Request bulk or custom format
Price (MYR)
688.00
Add to cart
Chemical Identifiers
CAS31558-41-5
IUPAC Name4-bromo-2,5-dimethoxybenzaldehyde
Molecular FormulaC9H9BrO3
InChI KeyBIFWGDWGCZLCHF-UHFFFAOYSA-N
SMILESCOC1=CC(Br)=C(OC)C=C1C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)>98.0%
Appearance (Color)White to yellow
FormCrystals or Crystalline powder
Free acid (titration)<1.0%
Paraffin wax, CnH2n+2, CAS Number-8002-74-2, propafenona inn-spanish, propafenona, propafenone-hcl, propafenonum inn-latin, propafenonum, propafenone inn:ban, propafenone inn, rythmol, propafenone, gnf-pf-4594, 2kg, 322.0 deg.C, CHEBI:63619, White, 232-315-6, 198 deg.C, Pure
RUO – Research Use Only