Octyl sulfate, sodium salt, 99%, HPLC grade, Thermo Scientific Chemicals
Octyl sulfate, sodium salt, 99%, HPLC grade, Thermo Scientific Chemicals
Octyl sulfate, sodium salt, 99%, HPLC grade, Thermo Scientific Chemicals
Octyl sulfate, sodium salt, 99%, HPLC grade, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Octyl sulfate, sodium salt, 99%, HPLC grade, Thermo Scientific Chemicals
CAS: 142-31-4 | C8H15Na·H2SO4 | 232.28 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number FSA416500250
Price (MYR)
4,093.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
4,093.00
Add to cart
Chemical Identifiers
CAS24358-62-1
IUPAC Name(1R)-1-(4-bromophenyl)ethan-1-aminium
Molecular FormulaC8H11BrN
InChI KeySOZMSEPDYJGBEK-ZCFIWIBFSA-O
SMILESC[C@@H]([NH3+])C1=CC=C(Br)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.5640-1.5700 @ 20?C
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥95.0%
for ion pair chromatography
RUO – Research Use Only