2-Hydroxy-5-nitrobenzaldehyde, 98+%, Thermo Scientific Chemicals
2-Hydroxy-5-nitrobenzaldehyde, 98+%, Thermo Scientific Chemicals
2-Hydroxy-5-nitrobenzaldehyde, 98+%, Thermo Scientific Chemicals
2-Hydroxy-5-nitrobenzaldehyde, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Hydroxy-5-nitrobenzaldehyde, 98+%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number FSA416180050
Price (MYR)
320.00
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
320.00
Add to cart
Chemical Identifiers
CAS1132-21-4
IUPAC Name3,5-dimethoxybenzoic acid
Molecular FormulaC9H10O4
InChI KeyIWPZKOJSYQZABD-UHFFFAOYSA-N
SMILESCOC1=CC(=CC(OC)=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point182°C to 185°C
Titration with NaOH97.5 to 102.5 %
HPLC>=97.5 %