4'-Nitroacetanilide, 99%, Thermo Scientific Chemicals
4'-Nitroacetanilide, 99%, Thermo Scientific Chemicals
4'-Nitroacetanilide, 99%, Thermo Scientific Chemicals
4'-Nitroacetanilide, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4'-Nitroacetanilide, 99%, Thermo Scientific Chemicals
CAS: 104-04-1 | C8H8N2O3 | 180.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number FSA415750250
Price (MYR)
784.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
784.00
Add to cart
Chemical Identifiers
CAS10340-91-7
IUPAC Name(isocyanomethyl)benzene
Molecular FormulaC8H7N
InChI KeyRIWNFZUWWRVGEU-UHFFFAOYSA-N
SMILES[C-]#[N+]CC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
AppearanceMay darken on storage
Refractive index1.5200 to 1.5220 (20°C, 589 nm)
Infrared spectrumConforms
GC>=97.5 %
Appearance (Color)Clear colorless to dark yellow