New methylene Blue N, pure
New methylene Blue N, pure
New methylene Blue N, pure
New methylene Blue N, pure
Thermo Scientific Chemicals
New methylene Blue N, pure
CAS: 1934-16-3 | C18H22ClN3S | 347.905 g/mol
Have Questions?
Catalog NumberQuantity
FSA4155300252.5 g
Catalog number FSA415530025
Price (MYR)
586.00
Add to cart
Quantity:
2.5 g
Request bulk or custom format
Price (MYR)
586.00
Add to cart
Chemical Identifiers
CAS10038-98-9
IUPAC Namegermanium tetrachloride
Molecular FormulaCl4GeH8
InChI KeyVDHBHZZBEGQJRK-UHFFFAOYSA-N
SMILESCl.Cl.Cl.Cl.[GeH4]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Appearance (Color)Clear colorless
Total trace metal impurities=<0.01 %