4-Fluoro-7-nitrobenzofurazan, 98%, Thermo Scientific Chemicals
4-Fluoro-7-nitrobenzofurazan, 98%, Thermo Scientific Chemicals
4-Fluoro-7-nitrobenzofurazan, 98%, Thermo Scientific Chemicals
4-Fluoro-7-nitrobenzofurazan, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Fluoro-7-nitrobenzofurazan, 98%, Thermo Scientific Chemicals
CAS: 29270-56-2 | C6H2FN3O3 | 183.1 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mg
25 mg
Packaging:
Glass bottle
Catalog number FSA415450050
Price (MYR)
931.00
Add to cart
Quantity:
5 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
931.00
Add to cart
Specifications
Chemical Name or Material4-Fluoro-7-nitrobenzofurazan
Name Note98%
CAS29270-56-2
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning