α-Naphtholbenzein
α-Naphtholbenzein
α-Naphtholbenzein
Thermo Scientific Chemicals
α-Naphtholbenzein
Indicator
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Catalog number FSA415320250
Price (MYR)
744.00
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
744.00
Add to cart
Chemical Identifiers
CAS579-74-8
IUPAC Name1-(2-methoxyphenyl)ethan-1-one
Molecular FormulaC9H10O2
InChI KeyDWPLEOPKBWNPQV-UHFFFAOYSA-N
SMILESCOC1=CC=CC=C1C(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.5380-1.5420 @ 20?C
Appearance (Color)Clear colorless to pale yellow
Identification (FTIR)Conforms
Assay (GC)≥97.5%