α-Naphtholbenzein
α-Naphtholbenzein
α-Naphtholbenzein
Thermo Scientific Chemicals
α-Naphtholbenzein
Indicator
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number FSA415320050
Price (MYR)
318.00
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
318.00
Add to cart
Specifications
Chemical Name or Materialα-Naphtholbenzein
CAS145-50-6
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee