Methylsulfuric acid sodium salt, 99%, pract., Thermo Scientific Chemicals
Methylsulfuric acid sodium salt, 99%, pract., Thermo Scientific Chemicals
Methylsulfuric acid sodium salt, 99%, pract., Thermo Scientific Chemicals
Methylsulfuric acid sodium salt, 99%, pract., Thermo Scientific Chemicals
Thermo Scientific Chemicals
Methylsulfuric acid sodium salt, 99%, pract., Thermo Scientific Chemicals
CAS: 512-42-5 | CH3NaO4S | 134.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA414900250
Price (MYR)
1,047.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,047.00
Add to cart
Chemical Identifiers
CAS7726-95-6
IUPAC Namebromide
Molecular FormulaBr
InChI KeyCPELXLSAUQHCOX-UHFFFAOYSA-M
SMILES[Br-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Orange to red to brown
Appearance (Form)Solution
Titration with Na2S2O30.9 to 1.5 M