Lactic acid, lithium salt, 99%, Thermo Scientific Chemicals
Lactic acid, lithium salt, 99%, Thermo Scientific Chemicals
Lactic acid, lithium salt, 99%, Thermo Scientific Chemicals
Lactic acid, lithium salt, 99%, Thermo Scientific Chemicals
Lactic acid, lithium salt, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Lactic acid, lithium salt, 99%, Thermo Scientific Chemicals
CAS: 867-55-0 | C3H5LiO3 | 96.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number FSA413335000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS103616-89-3
IUPAC Name(2S)-2-amino-3-(2-chlorophenyl)propanoic acid
Molecular FormulaC9H10ClNO2
InChI KeyCVZZNRXMDCOHBG-QMMMGPOBSA-N
SMILESN[C@@H](CC1=C(Cl)C=CC=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormPowder
Assay (HPLC)≥98.0%
Suitable for battery materials development.
RUO – Research Use Only