L-alpha-Lecithin, granular, from soybean oil
L-alpha-Lecithin, granular, from soybean oil
L-alpha-Lecithin, granular, from soybean oil
L-alpha-Lecithin, granular, from soybean oil
Thermo Scientific Chemicals
L-alpha-Lecithin, granular, from soybean oil
L-alpha-Lecithin, >-97% (phospholipids) min. (as acetone insoluble), C42H80NO8P, CAS Number-8002-43-5
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
Packaging:
Plastic bottle
Catalog number FSA413100010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS4052-92-0
IUPAC Name3-(chlorosulfonyl)benzoyl chloride
Molecular FormulaC7H4Cl2O3S
InChI KeyXWEBTVZIZWEJOO-UHFFFAOYSA-N
SMILESClC(=O)C1=CC=CC(=C1)S(Cl)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid after melting
Assay>=97.5 %
Infrared spectrumConforms
Refractive index1.5780 to 1.5800 (20°C, 589 nm)
Appearance (Color)Clear yellow