4-Isopropylbenzoic acid, 98+%, Thermo Scientific Chemicals
4-Isopropylbenzoic acid, 98+%, Thermo Scientific Chemicals
4-Isopropylbenzoic acid, 98+%, Thermo Scientific Chemicals
4-Isopropylbenzoic acid, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Isopropylbenzoic acid, 98+%, Thermo Scientific Chemicals
CAS: 536-66-3 | C10H12O2 | 164.2 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number FSA412800250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS16940-66-2
IUPAC Namesodium boranuide
Molecular FormulaBH4Na
InChI KeyYOQDYZUWIQVZSF-UHFFFAOYSA-N
SMILES[BH4-].[Na+]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Granules
Appearance (Color)White
Titration Iodometric>=97.5 %