N-Isopropylacrylamide, 99%, pure, stabilized
N-Isopropylacrylamide, 99%, pure, stabilized
N-Isopropylacrylamide, 99%, pure, stabilized
N-Isopropylacrylamide, 99%, pure, stabilized
Thermo Scientific Chemicals
N-Isopropylacrylamide, 99%, pure, stabilized
CAS: 2210-25-5 | C6H11NO | 113.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number FSA412781000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS565-67-3
IUPAC Name2-methylpentan-3-ol
Molecular FormulaC6H14O
InChI KeyISTJMQSHILQAEC-UHFFFAOYNA-N
SMILESCCC(O)C(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥97.0%
Refractive Index1.4145-1.4195 @ 20?C