2-Methyl-1-propanol, for HPLC
2-Methyl-1-propanol, for HPLC
2-Methyl-1-propanol, for HPLC
2-Methyl-1-propanol, for HPLC
Thermo Scientific Chemicals
2-Methyl-1-propanol, for HPLC
CAS: 78-83-1 | C4H10O | 74.12 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA4126400101 LGlass bottle
Catalog number FSA412640010
Price (MYR)
612.00
Add to cart
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
612.00
Add to cart
Chemical Identifiers
CAS7440-50-8
IUPAC Namecopper
Molecular FormulaCu
InChI KeyRYGMFSIKBFXOCR-UHFFFAOYSA-N
SMILES[Cu]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.001% max.
For HPLC
RUO – Research Use Only