Hydroxypropyl methacrylate, 97+%, mixture of isomers, stabilized, Thermo Scientific Chemicals
Hydroxypropyl methacrylate, 97+%, mixture of isomers, stabilized, Thermo Scientific Chemicals
Hydroxypropyl methacrylate, 97+%, mixture of isomers, stabilized, Thermo Scientific Chemicals
Hydroxypropyl methacrylate, 97+%, mixture of isomers, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Hydroxypropyl methacrylate, 97+%, mixture of isomers, stabilized, Thermo Scientific Chemicals
CAS: 27813-02-1 | C7H12O3 | 144.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
Packaging:
Glass bottle
Catalog number FSA412150010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialHydroxypropyl methacrylate
Name Note96.5%
CAS27813-02-1
CAS Min %97.0
Health Hazard 1GHS Signal Word: Warning