(S)-3-Hydroxy-3-phenylpropanoic acid, 99%, Thermo Scientific Chemicals
(S)-3-Hydroxy-3-phenylpropanoic acid, 99%, Thermo Scientific Chemicals
(S)-3-Hydroxy-3-phenylpropanoic acid, 99%, Thermo Scientific Chemicals
(S)-3-Hydroxy-3-phenylpropanoic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
(S)-3-Hydroxy-3-phenylpropanoic acid, 99%, Thermo Scientific Chemicals
CAS: 36567-72-3 | C9H10O3 | 166.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number FSA412120010
Price (MYR)
1,028.00
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,028.00
Add to cart
Specifications
Chemical Name or Material(S)-3-Hydroxy-3-phenylpropanoic acid
CAS36567-72-3
CAS Min %98.5
CAS Max %100.0
Melting Point116.0°C to 119.0°C