alpha-Hydroxyacetophenone, 97%
alpha-Hydroxyacetophenone, 97%
alpha-Hydroxyacetophenone, 97%
alpha-Hydroxyacetophenone, 97%
Thermo Scientific Chemicals
alpha-Hydroxyacetophenone, 97%
CAS: 582-24-1 | C8H8O2 | 136.15 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA41190025025 gGlass bottle
Catalog number FSA411900250
Price (MYR)
909.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
909.00
Add to cart
Chemical Identifiers
CAS3389-21-7
IUPAC Name3-(2-bromoethyl)-1H-indole
Molecular FormulaC10H10BrN
InChI KeyNTLAICDKHHQUGC-UHFFFAOYSA-N
SMILESBrCCC1=CNC2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Off-white to light brown
Infrared spectrumConforms
Melting point95°C to 101°C
Appearance (Form)Crystalline powder
NMR (1H-NMR)Consistent with structure