n-Heptane, 99+%, for spectroscopy, Thermo Scientific Chemicals
n-Heptane, 99+%, for spectroscopy, Thermo Scientific Chemicals
n-Heptane, 99+%, for spectroscopy, Thermo Scientific Chemicals
n-Heptane, 99+%, for spectroscopy, Thermo Scientific Chemicals
Thermo Scientific Chemicals
n-Heptane, 99+%, for spectroscopy, Thermo Scientific Chemicals
CAS: 142-82-5 | C7H16 | 100.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
500 mL
Packaging:
View all
Glass Bottle
Catalog number FSA411250025
Price (MYR)
1,476.00
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
1,476.00
Add to cart
Chemical Identifiers
CAS615-28-1
IUPAC Namedihydrogen benzene-1,2-diamine dichloride
Molecular FormulaC6H10Cl2N2
InChI KeyRIIWUGSYXOBDMC-UHFFFAOYSA-N
SMILES[H+].[H+].[Cl-].[Cl-].NC1=CC=CC=C1N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Appearance (Color)Light pink to purple
Titration Argentometric>=99.0 %
Infrared spectrumConforms
For UV/VIS spectroscopy
RUO – Research Use Only