Gibberellic acid, 99%, Thermo Scientific Chemicals
Gibberellic acid, 99%, Thermo Scientific Chemicals
Gibberellic acid, 99%, Thermo Scientific Chemicals
Gibberellic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Gibberellic acid, 99%, Thermo Scientific Chemicals
CAS: 77-06-5 | C19H22O6 | 346.38 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Packaging:
Glass bottle
Catalog number FSA410910050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS61379-64-4
IUPAC Name4-cyclopentylpiperazin-1-amine
Molecular FormulaC9H19N3
InChI KeyQYHRIASMJNLWHJ-UHFFFAOYSA-N
SMILESNN1CCN(CC1)C1CCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)>96.0%