Gallic Acid 98%, Thermo Scientific Chemicals
Gallic Acid 98%, Thermo Scientific Chemicals
Gallic Acid 98%, Thermo Scientific Chemicals
Gallic Acid 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Gallic Acid 98%, Thermo Scientific Chemicals
CAS: 149-91-7 | C7H6O5 | 170.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5 g
250 g
Packaging:
View all
Plastic bottle
Catalog number FSA410860010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS1118-02-1
IUPAC Nameisocyanatotrimethylsilane
Molecular FormulaC4H9NOSi
InChI KeyNIZHERJWXFHGGU-UHFFFAOYSA-N
SMILESC[Si](C)(C)N=C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.3925-1.3995 @ 20?C
FormLiquid
Appearance (Color)Clear to turbid colorless
Identification (FTIR)Conforms
Assay (GC)≥92.5%