Formic acid, Reagent ACS 88%, Thermo Scientific Chemicals
Formic acid, Reagent ACS 88%, Thermo Scientific Chemicals
Formic acid, Reagent ACS 88%, Thermo Scientific Chemicals
Formic acid, Reagent ACS 88%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Formic acid, Reagent ACS 88%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
500 mL
Packaging:
Plastic bottle
Catalog number FSA410770025
Price (MYR)
1,155.00
Add to cart
Quantity:
2.5 L
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
1,155.00
Add to cart
Chemical Identifiers
CAS141942-85-0
IUPAC Nameethyl (3R)-4-cyano-3-hydroxybutanoate
Molecular FormulaC7H11NO3
InChI KeyLOQFROBMBSKWQY-ZCFIWIBFSA-N
SMILESCCOC(=O)C[C@H](O)CC#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear, colourless to pale yellow
Assay (GC)> 97.5%
Refractive Index1.4460 - 1.4500 @20?C
Formliquid
Optical Rotation-4.5? ? 0.5?, neat