Fluosilicic acid, 25 wt.% aqueous solution, Thermo Scientific Chemicals
Fluosilicic acid, 25 wt.% aqueous solution, Thermo Scientific Chemicals
Fluosilicic acid, 25 wt.% aqueous solution, Thermo Scientific Chemicals
Fluosilicic acid, 25 wt.% aqueous solution, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Fluosilicic acid, 25 wt.% aqueous solution, Thermo Scientific Chemicals
CAS: 16961-83-4 | F6H2Si | 144.091 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
500 g
Packaging:
Plastic bottle
Catalog number FSA410710025
Price (MYR)
1,218.00
Add to cart
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
1,218.00
Add to cart
Chemical Identifiers
CAS2550-26-7
IUPAC Name4-phenylbutan-2-one
Molecular FormulaC10H12O
InChI KeyAKGGYBADQZYZPD-UHFFFAOYSA-N
SMILESCC(=O)CCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.5100 to 1.5130 (20°C, 589 nm)
Appearance (Form)Liquid
GC>=98.5 %
Specific gravity(25/25°C) 0.985 to 0.99
Appearance (Color)Clear colorless to light yellow