2-Ethylhexyl acetate, 99%, pure
2-Ethylhexyl acetate, 99%, pure
2-Ethylhexyl acetate, 99%, pure
2-Ethylhexyl acetate, 99%, pure
Thermo Scientific Chemicals
2-Ethylhexyl acetate, 99%, pure
CAS: 103-09-3 | C10H20O2 | 172.27 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA410105000500 gGlass bottle
Catalog number FSA410105000
Price (MYR)
260.00
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
260.00
Add to cart
Specifications
Chemical Name or Material2-Ethylhexyl acetate
Name Note99%
CAS103-09-3
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning