Ethyl 3-bromopropionate, 99%, Thermo Scientific Chemicals
Ethyl 3-bromopropionate, 99%, Thermo Scientific Chemicals
Ethyl 3-bromopropionate, 99%, Thermo Scientific Chemicals
Ethyl 3-bromopropionate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Ethyl 3-bromopropionate, 99%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Catalog number FSA409810250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS4798-44-1
IUPAC Namehex-1-en-3-ol
Molecular FormulaC6H12O
InChI KeyBVOSSZSHBZQJOI-UHFFFAOYNA-N
SMILESCCCC(O)C=C
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
Refractive Index1.4275-1.4315 @ 20?C
Appearance (Color)Clear colorless