1,4-Dioxane, 99+%, ACS reagent, stabilized, meets the requirements of Reag.Ph.Eur., Thermo Scientific Chemicals
1,4-Dioxane, 99+%, ACS reagent, stabilized, meets the requirements of Reag.Ph.Eur., Thermo Scientific Chemicals
1,4-Dioxane, 99+%, ACS reagent, stabilized, meets the requirements of Reag.Ph.Eur., Thermo Scientific Chemicals
1,4-Dioxane, 99+%, ACS reagent, stabilized, meets the requirements of Reag.Ph.Eur., Thermo Scientific Chemicals
Thermo Scientific Chemicals
1,4-Dioxane, 99+%, ACS reagent, stabilized, meets the requirements of Reag.Ph.Eur., Thermo Scientific Chemicals
CAS: 123-91-1 | C4H8O2 | 88.106 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
500 mL
2.5 L
Packaging:
View all
Glass Bottle
Catalog number FSA408820010
Price (MYR)
632.00
Add to cart
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
632.00
Add to cart
Chemical Identifiers
CAS123-92-2
IUPAC Name3-methylbutyl acetate
Molecular FormulaC7H14O2
InChI KeyMLFHJEHSLIIPHL-UHFFFAOYSA-N
SMILESCC(C)CCOC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Refractive index1.3990 to 1.4030 (20°C, 589 nm)
GC>=98.5 % (sum of isomers)
Appearance (Color)Clear colorless
Appearance (Form)Liquid