Dimethylglyoxime, 99+%, ACS reagent, Thermo Scientific Chemicals
Dimethylglyoxime, 99+%, ACS reagent, Thermo Scientific Chemicals
Dimethylglyoxime, 99+%, ACS reagent, Thermo Scientific Chemicals
Dimethylglyoxime, 99+%, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Dimethylglyoxime, 99+%, ACS reagent, Thermo Scientific Chemicals
CAS: 95-45-4 | C4H8N2O2 | 116.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
Packaging:
Plastic bottle
Catalog number FSA408331000
Price (MYR)
398.00
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
398.00
Add to cart
Chemical Identifiers
CAS62625-21-2
IUPAC Namesodium 2-{[4-hydroxy-2-methyl-5-(propan-2-yl)phenyl][(1E)-2-methyl-4-oxo-5-(propan-2-yl)cyclohexa-2,5-dien-1-ylidene]methyl}benzene-1-sulfonate
Molecular FormulaC27H29NaO5S
InChI KeyOCMIKNBSIRSUPI-FRWNMSGJSA-M
SMILES[Na+].CC(C)C1=C\C(=C(\C2=CC(C(C)C)=C(O)C=C2C)C2=C(C=CC=C2)S([O-])(=O)=O)C(C)=CC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Color ChangeVisual transition: pH 8.0 (Yellow) - pH 9.6 (Blue)
lambda-max (UV-Vis)593 - 601nm (0.01M NaOH)
FormPowder
Specific Absorbance (E 1%/1cm)≥ 550 at Lambda max
Appearance (Color)Orange to Dark Orange to Red-Brown to Dark green-Dark Brown