N,N-Dimethylcyclohexylamine, 99%, Thermo Scientific Chemicals
N,N-Dimethylcyclohexylamine, 99%, Thermo Scientific Chemicals
N,N-Dimethylcyclohexylamine, 99%, Thermo Scientific Chemicals
N,N-Dimethylcyclohexylamine, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
N,N-Dimethylcyclohexylamine, 99%, Thermo Scientific Chemicals
CAS: 98-94-2 | C8H17N | 127.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
Packaging:
Glass bottle
Catalog number FSA408255000
Price (MYR)
456.00
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
456.00
Add to cart
Chemical Identifiers
CAS298-83-9
IUPAC Name2-{3,3'-dimethoxy-4'-[3-(4-nitrophenyl)-5-phenyl-3H-1,2λ⁵,3,4-tetrazol-2-ylium-2-yl]-[1,1'-biphenyl]-4-yl}-3-(4-nitrophenyl)-5-phenyl-3H-1,2λ⁵,3,4-tetrazol-2-ylium dichloride
Molecular FormulaC40H30Cl2N10O6
InChI KeyFSVCQIDHPKZJSO-UHFFFAOYSA-L
SMILES[Cl-].[Cl-].COC1=CC(=CC=C1[N+]1=NC(=NN1C1=CC=C(C=C1)[N+]([O-])=O)C1=CC=CC=C1)C1=CC=C(C(OC)=C1)[N+]1=NC(=NN1C1=CC=C(C=C1)[N+]([O-])=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Loss on drying=<10 % (105°C)
Appearance (Form)Powder
HPLC>=88 %
Sulfated ash=<0.1 %