Diisopropanolamine, 99%, Thermo Scientific Chemicals
Diisopropanolamine, 99%, Thermo Scientific Chemicals
Diisopropanolamine, 99%, Thermo Scientific Chemicals
Diisopropanolamine, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Diisopropanolamine, 99%, Thermo Scientific Chemicals
CAS: 110-97-4 | C6H15NO2 | 133.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
Packaging:
Glass bottle
Catalog number FSA407805000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS26023-84-7
IUPAC Namedihydrogen hexachloroplatinumtetrakis(ylium)
Molecular FormulaCl6H2Pt
InChI KeyZKOQTCCVSRSECD-UHFFFAOYSA-J
SMILES[H+].[H+].Cl[Pt+4](Cl)(Cl)(Cl)(Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Orange to orange-brown
Total trace metal impurities=<0.1 %
Solubility(1 g in 10 ml ethanol) Clear orange
Appearance (Form)Powder and/or chunks
Assay38 to 40 % (Pt)