Diethylenetriaminepentaacetic acid, pentasodium salt, tech., 40% aqueous solution
Diethylenetriaminepentaacetic acid, pentasodium salt, tech., 40% aqueous solution
Diethylenetriaminepentaacetic acid, pentasodium salt, tech., 40% aqueous solution
Diethylenetriaminepentaacetic acid, pentasodium salt, tech., 40% aqueous solution
Thermo Scientific Chemicals
Diethylenetriaminepentaacetic acid, pentasodium salt, tech., 40% aqueous solution
CAS: 140-01-2 | C14H18N3Na5O10 | 503.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
3 kg
Packaging:
Glass bottle
Catalog number FSA407292500
Price (MYR)
290.00
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
290.00
Add to cart
Chemical Identifiers
CAS433-27-2
IUPAC Name1-ethoxy-2,2,2-trifluoroethan-1-ol
Molecular FormulaC4H7F3O2
InChI KeyKLXJPQNHFFMLIG-UHFFFAOYNA-N
SMILESCCOC(O)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=88 %
Appearance (Color)Clear colorless
Infrared spectrumConforms
Refractive index1.3410 to 1.3440 (20°C, 589 nm)
Chelation/Complexation reagent
RUO – Research Use Only