Diethylenetriaminepentaacetic acid, pentasodium salt, tech., 40% aqueous solution
Diethylenetriaminepentaacetic acid, pentasodium salt, tech., 40% aqueous solution
Diethylenetriaminepentaacetic acid, pentasodium salt, tech., 40% aqueous solution
Diethylenetriaminepentaacetic acid, pentasodium salt, tech., 40% aqueous solution
Thermo Scientific Chemicals
Diethylenetriaminepentaacetic acid, pentasodium salt, tech., 40% aqueous solution
CAS: 140-01-2 | C14H18N3Na5O10 | 503.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
3 kg
Packaging:
Catalog number FSA407290030
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Request bulk or custom format
Chemical Identifiers
CAS7758-09-0
IUPAC Namepotassium nitrite
Molecular FormulaKNO2
InChI KeyBXNHTSHTPBPRFX-UHFFFAOYSA-M
SMILES[K+].[O-]N=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with KMnO4>=96.0 %
Magnesium (Mg)=<20 ppm
Appearance (Color)White to light yellow
Heavy metals (as Pb)=<10 ppm
Sulfate (SO4)=<100 ppm
View more
Chelation/Complexation reagent
RUO – Research Use Only