Dichloromethane, 99.6%, ACS reagent, stabilized with amylene, Thermo Scientific Chemicals
Dichloromethane, 99.6%, ACS reagent, stabilized with amylene, Thermo Scientific Chemicals
Dichloromethane, 99.6%, ACS reagent, stabilized with amylene, Thermo Scientific Chemicals
Dichloromethane, 99.6%, ACS reagent, stabilized with amylene, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Dichloromethane, 99.6%, ACS reagent, stabilized with amylene, Thermo Scientific Chemicals
CAS: 75-09-2 | CH2Cl2 | 84.93 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
Catalog number FSA406920010
Price (MYR)
359.00
Add to cart
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
359.00
Add to cart
Specifications
Chemical Name or MaterialDichloromethane
Melting Point-97.0°C
Name NoteStabilized with Amylene
CAS75-09-2
Health Hazard 1Warning