4,4'-Diaminostilbene-2,2'-disulfonic acid, 95%, Thermo Scientific Chemicals
4,4'-Diaminostilbene-2,2'-disulfonic acid, 95%, Thermo Scientific Chemicals
4,4'-Diaminostilbene-2,2'-disulfonic acid, 95%, Thermo Scientific Chemicals
4,4'-Diaminostilbene-2,2'-disulfonic acid, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4,4'-Diaminostilbene-2,2'-disulfonic acid, 95%, Thermo Scientific Chemicals
CAS: 81-11-8 | C14H14N2O6S2 | 370.4 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Glass bottle
Catalog number FSA406415000
Price (MYR)
869.00
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
869.00
Add to cart
Chemical Identifiers
CAS612-82-8
IUPAC Namedihydrogen 3,3'-dimethyl-[1,1'-biphenyl]-4,4'-diamine dichloride
Molecular FormulaC14H18Cl2N2
InChI KeyLUKPNZHXJRJBAN-UHFFFAOYSA-N
SMILES[H+].[H+].[Cl-].[Cl-].CC1=CC(=CC=C1N)C1=CC=C(N)C(C)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Light beige to light grey
Infrared spectrumConforms
Appearance (Form)Crystalline powder
Titration Argentometric>=98.5 % (on dry substance)
Water=<5 % (K.F.)